Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human mGluR8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5277SP
This item is not returnable.
View return policy
Description
mGluR8 Monoclonal specifically detects mGluR8 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
mGluR8 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
O00222 | |
GRM8 | |
Chinese hamster ovary cell line CHO-derived recombinant human mGluR8 Gln34-Ser514 (predicted) Accession # O00222 | |
25 μg | |
Primary | |
Detects human mGluR8 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human mGluR1, 2, 3, 4, 5, or 7 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
476410 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ41058, glutamate metabotropic receptor 8, glutamate receptor, metabotropic 8, GPRC1H, GPRC1HMGLUR8GLUR8, GRM8, metabotropic glutamate receptor 8, MGC126724, mGlu8, mGluR8 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2918 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction