Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human mGluR8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5277
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5277 Supplier R&D Systems Supplier No. MAB5277
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

mGluR8 Monoclonal specifically detects mGluR8 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen mGluR8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 476410
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00222
Gene Alias FLJ41058, glutamate metabotropic receptor 8, glutamate receptor, metabotropic 8, GPRC1H, GPRC1HMGLUR8GLUR8, GRM8, metabotropic glutamate receptor 8, MGC126724, mGlu8, mGluR8
Gene Symbols GRM8
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human mGluR8 Gln34-Ser514 (predicted) Accession # O00222
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2918
Test Specificity Detects human mGluR8 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human mGluR1, 2, 3, 4, 5, or 7 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.