Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MICA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB13002500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB13002500 500 μg
MAB13002SP 25 μg
MAB13002100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB13002500 Supplier R&D Systems Supplier No. MAB13002500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MICA Monoclonal specifically detects MICA in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen MICA
Applications Western Blot, ELISA
Classification Monoclonal
Clone 159233
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q29983
Gene Alias FLJ60820, MGC111087, PERB11.1
Gene Symbols MICA
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human MICA Accession # Q29983
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507436
Test Specificity Detects human MICA in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MICB is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.