Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICA APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
$218.00 - $546.00
Specifications
Antigen | MICA |
---|---|
Clone | 159227 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1300A025
![]() |
R&D Systems
FAB1300A025 |
25 Tests |
Each for $218.00
|
|
|||||
FAB1300A100
![]() |
R&D Systems
FAB1300A100 |
100 Tests |
Each for $546.00
|
|
|||||
Description
MICA Monoclonal specifically detects MICA in Human samples. It is validated for Flow Cytometry.Specifications
MICA | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
100507436 | |
BaF3 mouse pro-B cell line transfected with human MICA | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
159227 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
FLJ60820, MGC111087, PERB11.1 | |
MICA | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human MICA in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human MICB. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title