Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICA/B Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB13001U100UG
This item is not returnable.
View return policy
Description
MICA/B Monoclonal specifically detects MICA/B in Human samples. It is validated for Flow Cytometry.Specifications
MICA/B | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q29983 | |
MICA | |
BaF3 mouse pro-B cell line transfected with human MICA Accession # Q29983 | |
RUO | |
100507436 | |
Human | |
IgG2a |
Flow Cytometry | |
159207 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2 | |
Mouse | |
100 μg | |
Primary | |
Detects human MICA/B in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction