Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICB Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB1599N025
This item is not returnable.
View return policy
Description
MICB Monoclonal specifically detects MICB in Human samples. It is validated for Flow Cytometry.Specifications
MICB | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4277 | |
Human | |
Purified |
Flow Cytometry | |
236511 | |
Flow Cytometry 5 uL/10^6 cells | |
CAI18747 | |
MICB | |
Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747 | |
25 Tests | |
Primary | |
Detects human MICB in direct ELISAs and Western blots. Does not cross-react with recombinant human MICA. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction