Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICB Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems BAF1599
This item is not returnable.
View return policy
Description
MICB Polyclonal specifically detects MICB in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
MICB | |
Polyclonal | |
Biotin | |
CAI18747 | |
MICB | |
Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747 | |
50 μg | |
Primary | |
Detects human MICB in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) MICA is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
Goat | |
Affinity Purified | |
RUO | |
4277 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction