Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MICB PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 2 publications

$198.10 - $467.50

Specifications

Antigen MICB
Clone 236511
Dilution Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB1599P025
GREENER_CHOICE
View Documents
R&D Systems
FAB1599P025
25 Tests
Each for $198.10
Only null left
Add to Cart
 
FAB1599P100
GREENER_CHOICE
View Documents
R&D Systems
FAB1599P100
100 Tests
Each for $467.50
Only null left
Add to Cart
 
Description

Description

MICB Monoclonal specifically detects MICB in Human samples. It is validated for Flow Cytometry, Knockout Validated.
Specifications

Specifications

MICB
Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog
MICB
IgG2b
Protein A or G purified from hybridoma culture supernatant
Detects human MICB in direct ELISAs and Western blots. Does not cross-react with recombinant human MICA.
236511
Flow Cytometry, Immunoassay
R-PE
Mouse
Human
CAI18747
4277
Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.