Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MICL/CLEC12A Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB29461V10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB29461V10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB29461V10 Supplier R&D Systems Supplier No. FAB29461V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MICL/CLEC12A Monoclonal specifically detects MICL/CLEC12A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MICL/CLEC12A
Applications Flow Cytometry
Classification Monoclonal
Clone 2439B
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q5QGZ9
Gene Alias CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A, Thr67-Ala265, Accession # Q5QGZ9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 160364
Test Specificity Detects human MICL/CLEC12A in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.