Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MICL/CLEC12A Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2946T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2946T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2946T100 Supplier R&D Systems Supplier No. FAB2946T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MICL/CLEC12A Monoclonal specifically detects MICL/CLEC12A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MICL/CLEC12A
Applications Flow Cytometry
Classification Monoclonal
Clone 687317
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q5QGZ9
Gene Alias CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Gene Symbols CLEC12A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A Thr67-Ala265 Accession # Q5QGZ9
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160364
Test Specificity Detects human MICL/CLEC12A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CLEC1, 2, 2A, 3B, 9A, 10A, 12B, 14A, rhCD302/CLEC13A recombinant mouse (rm) CLEC4B2, or rmMICL is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.