Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MICL/CLEC12A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2946
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2946 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2946 Supplier R&D Systems Supplier No. MAB2946
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MICL/CLEC12A Monoclonal specifically detects MICL/CLEC12A in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen MICL/CLEC12A
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 687317
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q5QGZ9
Gene Alias CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Gene Symbols CLEC12A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A Thr67-Ala265 Accession # Q5QGZ9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160364
Test Specificity Detects human MICL/CLEC12A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CLEC1, 2, 2A, 3B, 9A, 10A, 12B, 14A, rhCD302/CLEC13A recombinant mouse (rm) CLEC4B2, or rmMICL is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.