Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Midkine Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC2581N100UG
This item is not returnable.
View return policy
Description
Midkine Monoclonal specifically detects Midkine in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Midkine | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
E. coli-derived recombinant human Midkine., Lys23-Asp143, Accession No. P21741 | |
100 μg | |
4192.0 | |
Human | |
Supernatant |
Flow Cytometry | |
2284A | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
P21741 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Midkine in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction