Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Midkine Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2581SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2581SP 25 μg
MAB2581100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2581SP Supplier R&D Systems Supplier No. MAB2581SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Midkine Monoclonal specifically detects Midkine in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen Midkine
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2284A
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P21741
Gene Alias Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein
Host Species Rabbit
Immunogen E. coli-derived recombinant human Midkine. Lys23-Asp143 Accession # P21741
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 4192
Test Specificity Detects human Midkine in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.