Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Midkine Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $467.50

Specifications

Antigen Midkine
Clone 1011522
Dilution Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 1 ug/mL, Immunocytochemistry 5-25 ug/mL
Applications Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2582SP
GREENER_CHOICE
View Documents
R&D Systems
MAB2582SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB2582100
GREENER_CHOICE
View Documents
R&D Systems
MAB2582100
100 μg
Each for $467.50
Only null left
Add to Cart
 
Description

Description

Midkine Monoclonal specifically detects Midkine in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

Midkine
Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 1 ug/mL, Immunocytochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein
E. coli-derived recombinant human Midkine, Lys23-Asp143, Accession # P21741
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Midkine in direct ELISAs.
1011522
Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Unconjugated
Mouse
Human
P21741
4192
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.