Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Midkine Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
| Antigen | Midkine |
|---|---|
| Clone | 1011622 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 1 ug/mL |
| Applications | Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2583SP
![]() |
R&D Systems
MAB2583SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2583100
![]() |
R&D Systems
MAB2583100 |
100 μg |
Each for $432.50
|
|
|||||
Description
Midkine Monoclonal specifically detects Midkine in Human samples. It is validated for Immunohistochemistry.Specifications
| Midkine | |
| Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
| E. coli-derived recombinant human Midkine, Lys23-Asp143, Accession # P21741 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1011622 | |
| Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P21741 | |
| 4192 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title