Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Midkine Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF258
This item is not returnable.
View return policy
Description
Midkine Polyclonal specifically detects Midkine in Human samples. It is validated for Western Blot.Specifications
Midkine | |
Polyclonal | |
Biotin | |
P21741 | |
MDK | |
E. coli-derived recombinant human Midkine (R&D Systems, Catalog # 258-MD) Lys23-Asp143 Accession # P21741 | |
50 μg | |
Primary | |
Detects human Midkine in Western blots. In this format, less than 1% cross-reactivity with recombinant human Pleiotrophin is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
Goat | |
Affinity Purified | |
RUO | |
4192 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction