Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Mimecan Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2660SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2660SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2660SP Supplier R&D Systems Supplier No. AF2660SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Mimecan/OIF Polyclonal specifically detects Mimecan/OIF in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Mimecan/OIF
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P20774
Gene Alias DKFZp586P2421, KSPG25, mimecan, mimecan proteoglycan, OGN, OIF, OIFOG, osteoglycin, osteoglycin (osteoinductive factor), Osteoinductive factor, SLRR3A, SLRR3Acorneal keratan sulfate proteoglycan
Gene Symbols OGN
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Mimecan Ala20-Phe298 Accession # P20774
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4969
Test Specificity Detects human Mimecan in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant mouse Mimecan is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.