Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mineralocorticoid R/NR3C2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems PPH312200
This item is not returnable.
View return policy
Description
Mineralocorticoid R/NR3C2 Monoclonal specifically detects Mineralocorticoid R/NR3C2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA.Specifications
Mineralocorticoid R/NR3C2 | |
Monoclonal | |
Unconjugated | |
A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
NR3C2 | |
Recombinant human Mineralocorticoid R/NR3C2 aa 2-99 | |
100 μg | |
Primary | |
This antibody specifically recognizes human MR and cross-reacts with mouse and rat MR. Not yet tested in other species. | |
This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA | |
H3122 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA | |
FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MineralocorticoidR, MLR, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4306 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction