Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mineralocorticoid R/NR3C2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4369
This item is not returnable.
View return policy
Description
Mineralocorticoid R/NR3C2 Monoclonal specifically detects Mineralocorticoid R/NR3C2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mineralocorticoid R/NR3C2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
AAI11759 | |
NR3C2 | |
E. coli-derived recombinant human Mineralocorticoid R/NR3C2 Met1-Leu670 Accession # AAI11759 | |
100 μg | |
Primary | |
Detects human Mineralocorticoid R/NR3C2. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
385707 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MineralocorticoidR, MLR, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4306 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction