Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MIS/AMH Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB17372
This item is not returnable.
View return policy
Description
MIS/AMH Monoclonal specifically detects MIS/AMH in Human samples. It is validated for Western Blot.Specifications
MIS/AMH | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P03971 | |
AMH | |
Mouse myeloma cell line NS0-derived recombinant human MIS/AMH Leu19-Gln450 Accession # P03971 | |
100 μg | |
Primary | |
Detects human MIS/AMH in ELISAs. | |
Human | |
Purified |
Western Blot | |
805513 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
268 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction