Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MIS/AMH Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1737100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1737100 100 μg
MAB1737SP 25 μg
MAB1737500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1737100 Supplier R&D Systems Supplier No. MAB1737100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

MIS/AMH Monoclonal specifically detects MIS/AMH in Human samples. It is validated for Western Blot.

Specifications

Antigen MIS/AMH
Applications Western Blot
Classification Monoclonal
Clone 213816
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_000470
Gene Alias AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance
Gene Symbols AMH
Host Species Mouse
Immunogen E. coli-derived recombinant human MIS/AMH Ala453-Arg560 Accession # NP_000470
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268
Test Specificity Detects human MIS/AMH in direct ELISAs and Western blots. Shows approximately 50-100% cross-reactivity with recombinant mouse (rm) MIS and recombinant rat MIS and no cross-reactivity with recombinant human (rh) ART, rmArtemin, rhCripto-1, recombinant drosophila Dpp, rhGDNF, rhLAP, rhLatent TGF-beta 1, rhLefty-A (short and long isoforms), rmLefty-1 (short and long isoforms), rhNeurturin, rmNodal, rhTGF-alpha , recombinant amphibian TGF-beta 5, rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or rmWIF-1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.