Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MIS RII Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4749
This item is not returnable.
View return policy
Description
MIS RII/AMHR2 Polyclonal specifically detects MIS RII/AMHR2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
MIS RII/AMHR2 | |
Polyclonal | |
Unconjugated | |
Q16671 | |
AMHR2 | |
Mouse myeloma cell line NS0-derived recombinant human MIS RII Pro18-Ser144 Accession # Q16671 | |
100 μg | |
Primary | |
Detects human MIS RII in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant rat MIS RII is observed, and less than 1% cross-reactivity with recombinant human TGF-beta RI, RII, RIII, and RIIB is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
AMH type II receptor, AMHR2, AMHREC 2.7.11.30, AMHRII, C14, MISR2, MISRII, MISRIIMIS type II receptor, MRII, Muellerian hormone type II receptor, Muellerian hormone type-2 receptor, Mullerian hormone receptor, type II, Mullerian inhibiting substance type II receptor | |
Sheep | |
Affinity Purified | |
RUO | |
269 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction