Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MKK4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3390SP
This item is not returnable.
View return policy
Description
MKK4/MEK4 Monoclonal specifically detects MKK4/MEK4 in Human samples. It is validated for Immunocytochemistry.Specifications
MKK4/MEK4 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P45985 | |
MAP2K4 | |
E. coli-derived recombinant human MKK4 Met1-Asp399 Accession # P45985 | |
25 μg | |
Primary | |
Detects human MKK4 in direct ELISAs. This antibody does not cross-react with recombinant human MKK7. | |
Human | |
Purified |
Immunocytochemistry | |
323042 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-Jun N-terminal kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 4, JNK activating kinase 1, JNK-activated kinase 1, JNK-activating kinase 1, JNKK, JNKK1, JNKK1MAPK/ERK kinase 4, MAP kinase kinase 4, MAP2K4, MAPKK4, MEK4, MEK4MEK 4, mitogen-activated protein kinase kinase 4, MKK4SAPK/ERK kinase 1, PRKMK4, PRKMK4MAPKK 4, SEK1, SERK1, SERK1EC 2.7.12.2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6416 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction