Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MKK7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3579
This item is not returnable.
View return policy
Description
MKK7/MEK7 Monoclonal specifically detects MKK7/MEK7 in Human samples. It is validated for Immunohistochemistry.Specifications
MKK7/MEK7 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
O14733 | |
MAP2K7 | |
E. coli-derived recombinant human MKK7 Met1-Arg419 Accession # O14733 | |
100 μg | |
Primary | |
Detects rhMKK7 in direct ELISAs. This antibody does not cross-react with rhMKK4. | |
Human | |
Purified |
Immunohistochemistry | |
324302 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-Jun N-terminal kinase kinase 2, dual specificity mitogen-activated protein kinase kinase 7, EC 2.7.12.2, JNK kinase 2, JNK-activating kinase 2, JNKK 2, JNKK2, MAP kinase kinase 7, MAP2K7, MAPK/ERK kinase 7, MAPKK7, MEK 7, MEK7, mitogen-activated protein kinase kinase 7, MKE7, MKK7MAPKK 7, PRKMK7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5609 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction