Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MKP-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7168
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7168 Supplier R&D Systems Supplier No. MAB7168
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

MKP-1/DUSP1 Monoclonal specifically detects MKP-1/DUSP1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen MKP-1/DUSP1
Applications Immunohistochemistry
Classification Monoclonal
Clone 408612
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P28562
Gene Alias CL100, CL100MAP kinase phosphatase 1, dual specificity phosphatase 1, Dual specificity protein phosphatase hVH1, DUSP1, EC 3.1.3.16, EC 3.1.3.48, HVH1, Mitogen-activated protein kinase phosphatase 1, MKP1, MKP-1dual specificity protein phosphatase 1, Protein-tyrosine phosphatase CL100, PTPN10, PTPN10serine/threonine specific protein phosphatase, VH1
Gene Symbols DUSP1
Host Species Rat
Immunogen E. coli-derived recombinant human MKP-1 Met1-Cys367 Accession # P28562
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1843
Test Specificity Detects human MKP-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) MKP-2, rhMKP-3, or rhMKP-5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.