Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MKP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB35761100
This item is not returnable.
View return policy
Description
MKP-3/DUSP6 Monoclonal specifically detects MKP-3/DUSP6 in Human samples. It is validated for Immunocytochemistry.Specifications
MKP-3/DUSP6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
dual specificity phosphatase 6, Dual specificity protein phosphatase PYST1, DUSP6, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP3, MKP-3, MKP3serine/threonine specific protein phosphatase, PYST1, PYST1dual specificity protein phosphatase 6 | |
E. coli-derived recombinant human MKP-3, Met1-Thr381, Accession No. Q16828 | |
100 μg | |
1848.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Immunocytochemistry | |
363209 | |
Immunocytochemistry 8-25 ug/mL | |
Q16828 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human MKP-3 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction