Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MLK3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4559
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4559 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4559 Supplier R&D Systems Supplier No. AF4559
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

MLK3 Polyclonal specifically detects MLK3 in Human samples. It is validated for Western Blot.

Specifications

Antigen MLK3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q16584
Gene Alias EC 2.7.11, EC 2.7.11.25, MAP3K11, MEKK11, mitogen-activated protein kinase kinase kinase 11, Mixed lineage kinase 3, MLK3MLK-3, PTK1, PTK1protein-tyrosine kinase PTK1, RHOE, SH3 domain-containing proline-rich kinase, SPRK, SPRKMGC17114, Src-homology 3 domain-containing proline-rich kinase
Gene Symbols MAP3K11
Host Species Sheep
Immunogen E. coli-derived recombinant human MLK3 Ala85-Gly207 Accession # Q16584
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4296
Test Specificity Detects endogenous human MLK3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.