Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MLXIPL Partial ORF (NP_116569.1, 603 a.a. - 700 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7100345
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a basic helix-loop-helix leucine zipper transcription factor of the Myc/Max/Mad superfamily. This protein forms a heterodimeric complex and binds and activates, in a glucose-dependent manner, carbohydrate response element (ChoRE) motifs in the promoters of triglyceride synthesis genes. The gene is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11.23. [provided by RefSeq]

Sequence: PPAPSGSERRLSGDLSSMPGPGTLSVRVSPPQPILSRGRPDSNKTENRRITHISAEQKRRFNIKLGFDTLHGLVSTLSAQPSLKVSKATTLQKTAEYI

Specifications

Accession Number NP_116569.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51085
Molecular Weight (g/mol) 36.52kDa
Name MLXIPL (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PPAPSGSERRLSGDLSSMPGPGTLSVRVSPPQPILSRGRPDSNKTENRRITHISAEQKRRFNIKLGFDTLHGLVSTLSAQPSLKVSKATTLQKTAEYI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CHREBP/MIO/MONDOB/WBSCR14/WS-bHLH/bHLHd14
Common Name MLXIPL
Gene Symbol MLXIPL
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.