Learn More
Human MMP-1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | MMP-1 |
| Applications | Western Blot, Immunohistochemistry, KnockDown |
| Classification | Monoclonal |
| Clone | 36665R |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Knockout Validated |
| Formulation | Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P03956 |
| Gene Alias | CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1 |
| Gene Symbols | MMP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.