Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP-10 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC9102T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9102T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC9102T100 Supplier R&D Systems Supplier No. IC9102T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-10 Monoclonal specifically detects MMP-10 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen MMP-10
Applications Flow Cytometry
Classification Monoclonal
Clone 110316
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P09238
Gene Alias EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2
Gene Symbols MMP10
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMP-10 Phe99-Cys476 Accession # P09238
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4319
Test Specificity Detects the pro and active forms of human MMP-10 in direct ELISAs Western blots. In direct ELISAs and Western blots, approximately 10-50% cross-reactivity with recombinant human (rh) MMP-3 and no cross-reactivity with rhMMP-1, -2, -7, -8, -9, -12, or -13 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.