Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP-10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9102SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9102SP 25 μg
MAB9102 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9102SP Supplier R&D Systems Supplier No. MAB9102SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-10 Monoclonal specifically detects MMP-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen MMP-10
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 110316
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P09238
Gene Alias EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2
Gene Symbols MMP10
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMP-10 Phe99-Cys476 Accession # P09238
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4319
Test Specificity Detects the pro and active forms of human MMP-10 in direct ELISAs Western blots. In direct ELISAs and Western blots, approximately 10-50% cross-reactivity with recombinant human (rh) MMP-3 and no cross-reactivity with rhMMP-1, -2, -7, -8, -9, -12, or -13 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.