Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3657
This item is not returnable.
View return policy
Description
MMP-11 Monoclonal specifically detects MMP-11 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
MMP-11 | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P24347 | |
MMP11 | |
E. coli-derived recombinant human MMP-11 Met1-Leu488 Accession # P24347 | |
100 μg | |
Primary | |
Detects human MMP-11 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
135421 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.24, matrix metallopeptidase 11 (stromelysin 3), matrix metalloproteinase 11 (stromelysin 3), Matrix metalloproteinase-11, MMP11, MMP-11, SL-3, ST3, STMY3stromelysin 3, stromelysin III, stromelysin-3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4320 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction