Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP-13 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$214.00 - $2020.00

Specifications

Antigen MMP-13
Clone 87512
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification, Immunocytochemistry 8-25 ug/mL
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB511SP
GREENER_CHOICE
View Documents
R&D Systems
MAB511SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB511100
GREENER_CHOICE
View Documents
R&D Systems
MAB511100
100 μg
Each for $538.00
Only null left
Add to Cart
 
MAB511500
GREENER_CHOICE
View Documents
R&D Systems
MAB511500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

MMP-13 Monoclonal specifically detects MMP-13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification, Immunocytochemistry.
Specifications

Specifications

MMP-13
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification, Immunocytochemistry 8-25 ug/mL
Monoclonal
Mouse
Human
P45452
4322
Chinese hamster ovary cell line CHO-derived recombinant human MMP-13 Leu20-Cys471 Accession # P45452
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human MMP-13 in direct ELISAs and Western blots. Detects both pro and active forms of human MMP-13 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -12 is observed.
87512
Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification, Immunocytochemistry
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13
MMP13
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.