Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | MMP-13 |
---|---|
Clone | 87511 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5111SP
![]() |
R&D Systems
MAB5111SP |
25 μg |
Each for $200.50
|
|
|||||
MAB5111100
![]() |
R&D Systems
MAB5111100 |
100 μg |
Each for $432.50
|
|
|||||
Description
MMP-13 Monoclonal specifically detects MMP-13 in Human samples. It is validated for ELISA.Specifications
MMP-13 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P45452 | |
4322 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
87511 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13 | |
Chinese hamster ovary cell line CHO-derived recombinant human MMP-13, Leu20-Cys471, Accession # P45452 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human MMP-13 in direct ELISAs. In a sandwich immunoassay, detects human pro-MMP-13 when paired with an anti-human pro-MMP-13 detection antibody. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title