Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-13 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF511
This item is not returnable.
View return policy
Description
MMP-13 Polyclonal specifically detects MMP-13 in Human samples. It is validated for Western Blot.Specifications
MMP-13 | |
Polyclonal | |
Biotin | |
P45452 | |
MMP13 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-13 (R&D Systems, Catalog # 511-MM) Leu20-Cys471 Accession # P45452 | |
50 μg | |
Primary | |
Detects human MMP-13 in Western blots. In this format, approximately 5% cross-reactivity with recombinant human (rh) MMP-8 is observed and less than 2% cross-reactivity with rhMMP-1, -2, -3, -7, -9, and -12 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13 | |
Goat | |
Affinity Purified | |
RUO | |
4322 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction