Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-16/MT3-MMP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1785SP
This item is not returnable.
View return policy
Description
MMP-16/MT3-MMP Monoclonal specifically detects MMP-16/MT3-MMP in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| MMP-16/MT3-MMP | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P51512 | |
| MMP16 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-16/MT3-MMP Ala32-Pro535 Accession # P51512 | |
| 25 μg | |
| Primary | |
| Detects human MMP-16/MT3-MMP in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human MMP-14, -15, or -24 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 782005 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chromosome 8 open reading frame 57, DKFZp761D112, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 16 (membrane-inserted), matrix metalloproteinase 16 (membrane-inserted), matrix metalloproteinase-16, Membrane-type matrix metalloproteinase 3, Membrane-type-3 matrix metalloproteinase, MMP16, MMP-16, MMPX2, MMP-X2, MT3MMP, MT3-MMP, MT3-MMPC8orf57, MT-MMP 3, MT-MMP2, MTMMP3, MT-MMP3, Putative transmembrane protein C8orf57 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4325 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction