Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-17 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7796SP
This item is not returnable.
View return policy
Description
MMP-17 Monoclonal specifically detects MMP-17 in Human samples. It is validated for Neutralization.Specifications
MMP-17 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP17, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4326 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Neutralization | |
823511 | |
Neutralization <9 ug/mL | |
Q9ULZ9 | |
MMP17 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-17 Pro42-Ala534 Accession # Q9ULZ9 | |
25 μg | |
Primary | |
Detects human MMP-17 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human MMP-24 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction