Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP-17 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7796
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7796 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7796 Supplier R&D Systems Supplier No. MAB7796
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-17 Monoclonal specifically detects MMP-17 in Human samples. It is validated for Neutralization.

Specifications

Antigen MMP-17
Applications Neutralization
Classification Monoclonal
Clone 823511
Conjugate Unconjugated
Dilution Neutralization <9 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9ULZ9
Gene Alias matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP17, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4
Gene Symbols MMP17
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMP-17 Pro42-Ala534 Accession # Q9ULZ9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4326
Test Specificity Detects human MMP-17 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human MMP-24 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.