Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB905RSP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB905RSP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB905RSP Supplier R&D Systems Supplier No. MAB905RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Western Blot, Immunoaffinity Purification.

Specifications

Antigen MMP-3
Applications Western Blot, Affinity Purification
Classification Monoclonal
Clone 10D6R
Conjugate Unconjugated
Dilution Western Blot 1 μ/mL, Immunoaffinity Purification 25 μ/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP3, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, Stromelysin 1, stromelysin-1, Transin-1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human MMP-3
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 4314
Test Specificity Detects human MMP-3 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.