Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-8 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC9081R100UG
This item is not returnable.
View return policy
Description
MMP-8 Monoclonal specifically detects MMP-8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
MMP-8 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
AAZ38714 | |
MMP8 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-8 Phe21-Gly467 Accession # AAZ38714 | |
RUO | |
4317 | |
Human | |
IgG2a |
Flow Cytometry | |
100608 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CLG1HNC, Collagenase 2, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
Mouse | |
100 μg | |
Primary | |
Detects both pro and active forms of human MMP-8 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) MMP-1, -2, -3, -7, -9, -10, -12, or -13 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction