Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP-9 Antibody, R&D Systems™

Catalog No. MAB911SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB911SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB911SP Supplier R&D Systems Supplier No. MAB911SP
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 14 publications

MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, CyTOF-reported.

Specifications

Antigen MMP-9
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 4H3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 25-100 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase
Gene Symbols MMP9
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human MMP-9
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4318
Test Specificity Detects human MMP-9 in Western blots. In Western blots, reactivity with the pro (92 kDa), active (82 kDa), and C-terminal truncated (65 kDa) forms of recombinant human (rh) MMP-9 is observed. Also in Western blots, 20% cross-reactivity with rhMMP-2, 5% cross-reactivity with rhMMP-1, and no cross-reactivity with rhMMP-3, -7, -8, -10, -12, or -13 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.