Learn More
Human MMP-9 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB911RSP
This item is not returnable.
View return policy
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MMP-9 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human MMP-9 in direct ELISAs and Western blots. | |
Human | |
Supernatant |
Western Blot, Immunoprecipitation | |
4H3R | |
Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
Chinese hamster ovary cell line CHO-derived recombinant human MMP-9 | |
25 μg | |
4318 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |