Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human MMP-9 Quantikine ELISA Kit

Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Description
The Human MMP-9 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-9 in cell culture supernates (10 ul), serum (10 ul), platelet-poor heparin plasma (10 ul), saliva (10 ul), urine (100 ul).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Assay Range | 0.31 to 20 ng/mL |
Assay Sensitivity | 0.156 ng/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Platelet-poor Plasma (Heparin), Saliva, Serum, Urine |
Specificity | Natural and Recombinant Human 92 kDa Pro-MMP-9 and the 82 kDa active MMP-9 |
Sufficient For | 1 x 96-well Plate |
Target Species | Human |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction