Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMR/CD206 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB25342A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB25342A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB25342A Supplier R&D Systems Supplier No. FAB25342A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMR/CD206/Mannose Receptor Monoclonal specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MMR/CD206/Mannose Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone 685641
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P22897
Gene Alias CD206, CLEC13D, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen, MRC1
Gene Symbols MRC1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206 Leu19-Lys1383 (Thr399Ala) & (Leu407Phe) Accession # P22897
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4360
Test Specificity Detects human MMR/CD206 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse MMR is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.