Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse ADAMTS1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5867
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5867 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5867 Supplier R&D Systems Supplier No. BAF5867
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ADAMTS1 Polyclonal specifically detects ADAMTS1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen ADAMTS1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P97857
Gene Alias a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH1, METH-1
Gene Symbols ADAMTS1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse ADAMTS1 Phe254-Cys725 Accession # P97857
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9510
Test Specificity Detects human and mouse ADAMTS1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) ADAMTS4, rhADAMTSL1.2, and rhADAMTS15 is observed and less than 1% cross-reactivity with rhADAMTSL2, rhADAMTS5, rhADAMTS8, rhADAMTS10, rhADAMTS12, rhADAMTS13, and rhADAMTS16 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.