Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Adenosine Deaminase/ADA Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7048
This item is not returnable.
View return policy
Description
Adenosine Deaminase/ADA Polyclonal specifically detects Adenosine Deaminase/ADA in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.Specifications
Adenosine Deaminase/ADA | |
Polyclonal | |
Unconjugated | |
P00813 | |
ADA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Adenosine Deaminase/ADA Met1-Leu363 Accession # P00813 | |
100 μg | |
Primary | |
Detects human and mouse Adenosine Deaminase/ADA in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Knockout Validated | |
ADA, ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
Sheep | |
Affinity Purified | |
RUO | |
100 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction