Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Aldehyde Dehydrogenase 1-A1/ ALDH1A1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF5869
This item is not returnable.
View return policy
Description
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Polyclonal specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.Specifications
Aldehyde Dehydrogenase 1-A1/ALDH1A1 | |
Polyclonal | |
Unconjugated | |
P00352 | |
ALDH1A1 | |
E. coli-derived recombinant human Aldehyde Dehydrogenase 1-A1/ALDH1A1 Ser2-Ser501 Accession # P00352 | |
100 μg | |
Primary | |
Detects human Aldehyde Dehydrogenase 1-A1/ALDH1A1 in direct ELISAs and detects human and mouse Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde Dehydrogenase 1A1, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH1A1, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1 | |
Goat | |
Affinity Purified | |
RUO | |
216 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction