Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse AMPK beta 2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3808SP
This item is not returnable.
View return policy
Description
AMPK beta 2/PRKAB2 Monoclonal specifically detects AMPK beta 2/PRKAB2 in Human, Mouse samples. It is validated for Western Blot.Specifications
AMPK beta 2/PRKAB2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
O43741 | |
PRKAB2 | |
E. coli-derived recombinant human AMPK beta 2 Met1-Ile272 Accession # O43741 | |
25 μg | |
Primary | |
Detects human and mouse AMPK beta 2 in Western blots. No cross-reactivity with recombinant human AMPK beta 1 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
376938 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, PRKAB2, protein kinase, AMP-activated, beta 2 non-catalytic subunit | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5565 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction