Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse AMPK beta 2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3808
This item is not returnable.
View return policy
Description
AMPK beta 2/PRKAB2 Monoclonal specifically detects AMPK beta 2/PRKAB2 in Human, Mouse samples. It is validated for Western Blot.Specifications
AMPK beta 2/PRKAB2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
O43741 | |
PRKAB2 | |
E. coli-derived recombinant human AMPK beta 2 Met1-Ile272 Accession # O43741 | |
100 μg | |
Primary | |
Detects human and mouse AMPK beta 2 in Western blots. No cross-reactivity with recombinant human AMPK beta 1 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
376938 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, PRKAB2, protein kinase, AMP-activated, beta 2 non-catalytic subunit | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5565 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction