Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Angiopoietin-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB098100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB098100 100 μg
MAB098SP 25 μg
MAB098500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB098100 Supplier R&D Systems Supplier No. MAB098100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human, Mouse samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Angiopoietin-2
Applications Immunohistochemistry, ELISA
Classification Monoclonal
Clone 85816
Conjugate Unconjugated
Dilution Immunohistochemistry 1-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O15123
Gene Alias AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, ANGPT2, Tie2-ligand
Gene Symbols ANGPT2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-2 Asp68-Phe496 Accession # O15123
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285
Test Specificity Detects human and mouse Angiopoietin-2 in ELISAs. In ELISAs, no cross-reactivity or interference with recombinant human (rh) Ang-1, rhAng-4, rhAng-X, recombinant mouse (rm) Ang-3, rmANGPTL-3, rhTie-1, rhTie-2, and rmTie-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.